Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCL1, Mouse, Clone: MCL.C9, Abnova™

Catalog No. 89113602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-602 Supplier Abnova Corporation Supplier No. MAB8825
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MCL1.

The protein encoded by this gene belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing. [provided by RefSeq

Specifications

Antigen MCL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone MCL.C9
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MCL1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Gene MCL1
Gene Alias BCL2L3/EAT/MCL1L/MCL1S/MGC104264/MGC1839/TM
Gene Symbols MCL1
Host Species Mouse
Immunogen Recombinant protein corresponding to human MCL1.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4170
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.