Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Mcpip1, Clone: 2H8.2,

Catalog No. MABS430MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS430MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS430MI Supplier MilliporeSigma™ Supplier No. MABS430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Mcpip1 clone: 2H8.2 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)

MCP-induced protein 1 (also known as ribonuclease ZC3H12A, Zinc finger CCCH domain-containing protein 12A) is encoded by the ZC3H12A gene in human (also known as dJ423B22.1, MCPIP, MCPIP1). It belongs to the 4-member CCCH-zinc finger protein family (MCPIP1/2/3/4) that act as negative regulators in macrophage activation (PMID 18178554). MCPIP1 in particular is involved in immune response downregulation upon LPS treatment by targeting IL-6 and IL-12b with its RNase activity (PMID 22561375). MCPIP1 expresssion by MCP-1 treatment is reported to promote angiogenesis in human umbilical vein endothelial cell (HUVEC) cultures, while causing apoptotic cell death in cardiac myocytes (PMID 18364357 & 16574901).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mcpip1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H8.2
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q5D1E8
Gene Symbols ZC3H12A; MCPIP; MCPIP1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Mcpip1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_079355
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.