Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mdc1, Mouse, R-PE, Clone: P2B11, Abnova™

Catalog No. 89109628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-628 Supplier Abnova Corporation Supplier No. MAB11476
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant partial Mdc1.

Specifications

Antigen Mdc1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone P2B11
Conjugate R-PE
Description Mouse monoclonal antibody raised against recombinant partial Mdc1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Mdc1
Gene Alias 6820401C03/AA413496/Nfbd1/mKIAA0170
Gene Symbols Mdc1
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to C-terminus of mouse Mdc1.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240087
Target Species Bovine, Chimpanzee, Human, Mouse
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.