Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM4, Mouse, Clone: 15, Abnova™

Catalog No. 89113025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-025 Supplier Abnova Corporation Supplier No. MAB6760

Mouse monoclonal antibody raised against synthetic phosphopeptide of MDM4.

The human MDM4 gene, which plays a role in apoptosis, encodes a 490-amino acid protein containing a RING finger domain and a putative nuclear localization signal. The MDM4 putative nuclear localization signal, which all Mdm proteins contain, is located in the C-terminal region of the protein. The mRNA is expressed at a high level in thymus and at lower levels in all other tissues tested. MDM4 protein produced by in vitro translation interacts with p53 via a binding domain located in the N-terminal region of the MDM4 protein. MDM4 shows significant structural similarity to p53-binding protein MDM2. Two transcript variants, one protein-coding and the other likely not to be protein-coding, have been found for this gene. [provided by RefSeq

Specifications

Antigen MDM4
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone #15
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of MDM4.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene MDM4
Gene Alias DKFZp781B1423/HDMX/MDMX/MGC132766/MRP1
Gene Symbols MDM4
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S367 of human MDM4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4194
Test Specificity This antibody reacts with MDM4 phosphorylated at Ser367.
Target Species Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.