Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED8, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107533 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-533 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-533 Supplier Abnova Corporation Supplier No. H00112950A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MED8.

This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq

Sequence: MRQTEGRVPVFSHEVVPDHLRTKPDPEVEEQEKQLTTDAARIGADAAQKQIQSLNKMCS

Specifications

Antigen MED8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MED8.
Formulation 50% glycerol
Gene MED8
Gene Accession No. NM_052877
Gene Alias ARC32/MGC17544/MGC19641
Gene Symbols MED8
Host Species Mouse
Immunogen MED8 (NP_443109, 1 a.a. ∽ 59 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112950
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.