Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2B, Mouse, Clone: 4B5, Abnova™

Catalog No. 89104906 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-906 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-906 Supplier Abnova Corporation Supplier No. H00004207M24A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MEF2B.

This gene represents numerous read-through transcripts that span geneID:729991 and 100271849. Many read-through transcripts are predicted to be nonsense-mediated decay (NMD) candidates, and are thought to be noncoding. Some transcripts are predicted to be capable of translation reinitation at a downstream AUG, resulting in expression of at least one isoform of myocyte enhancer factor 2B (MEF2B) from this read-through locus. At least one additional MEF2B variant and isoform can be expressed from a downstream promoter, and is annotated on geneID:100271849. [provided by RefSeq

Sequence: SRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTEGRRSDLPGGLAGPRGGLNTSRSLYSGLQNP

Specifications

Antigen MEF2B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MEF2B.
Formulation ascites with no preservative
Gene MEF2B
Gene Accession No. NM_005919
Gene Alias FLJ32599/FLJ46391/MGC189732/MGC189763/RSRFR2
Gene Symbols MEF2B
Host Species Mouse
Immunogen MEF2B (NP_005910.1,165 a.a. ∽ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4207
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.