Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEMO1, Mouse, Clone: AT1E9, Abnova™

Catalog No. 89109364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-364 Supplier Abnova Corporation Supplier No. MAB11196
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MEMO1.

Specifications

Antigen MEMO1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1E9
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MEMO1.
Dilution Western Blot (1:3000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide.
Gene MEMO1
Gene Alias C2orf4/CGI-27/DKFZp434I0135/FLJ25031/MEMO/NS5ATP7
Gene Symbols MEMO1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-297 of human MEMO1.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51072
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.