Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Metallothionein, Clone: UC1MT, Abnova™

Catalog No. 89136450 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-450 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-136-450 Supplier Abnova Corporation Supplier No. MAB8275
This item is not returnable. View return policy

Specifications

Antigen Metallothionein
Applications Western Blot
Classification Monoclonal
Clone UC1MT
Conjugate Unconjugated
Formulation In PBS
Host Species Mouse
Immunogen Native purified rabbit Metallothionein.
Quantity 200 μg
Primary or Secondary Primary
Target Species Equine, Fish, Human, Mouse, Rabbit, Rat
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.