Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methicillin-resistant Staphylococcus aureus, Mouse, Clone: B483M, Abnova™

Catalog No. 89135672 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-672 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-672 Supplier Abnova Corporation Supplier No. MAB4260
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Methicillin-resistant Staphylococcus aureus.

Specifications

Antigen Methicillin-resistant Staphylococcus aureus
Applications ELISA, Western Blot
Classification Monoclonal
Clone B483M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Methicillin-resistant Staphylococcus aureus.
Dilution ELISA (1:5000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Mouse
Immunogen Purified Methicillin-resistant Staphylococcus aureus protein.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the product of the MRSA gene expressed by methicillin-resistant Staphylococcus aureus.
Target Species Staphylococcus species
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.