Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFI2, Mouse, Clone: 1F7, Abnova™

Catalog No. 89104919 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-919 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-919 Supplier Abnova Corporation Supplier No. H00004241M04A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MFI2.

The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants. [provided by RefSeq

Sequence: MEVRWCATSDPEQHKCGNMSEAFREAGIQPSLLCVRGTSADHCVQLIAAQEADAITLDGGAIYEAGKEHGLKPVVGEVYDQEVGTSYYAVAVVRRSSHVTIDTLKGVKSCHTGINRTVGWNVPVGYLVESGRLSVMGCDVLKAVSDHFGGSCVPGAGETSYSESLCRLCRGDSPGEGVCDKSPLERYYDYSGAFRCLAEGAGDVAFVKHSTVLENTDESPSRRQTWTRSEEEEGECPAHEEARRTMRSSAGQAWKWAPVHRPQDESDKGEFGKRAKSRDMLG

Specifications

Antigen MFI2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MFI2.
Formulation ascites with no preservative
Gene MFI2
Gene Accession No. BC001875
Gene Alias CD228/FLJ38863/MAP97/MGC4856/MTF1
Gene Symbols MFI2
Host Species Mouse
Immunogen MFI2 (AAH01875,21 a.a. ∽ 302 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4241
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.