Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGAT1, Mouse, Clone: 2G4, Abnova™

Catalog No. 89104920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-920 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-920 Supplier Abnova Corporation Supplier No. H00004245M08A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MGAT1.

There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: YLQREAYDRDFLARVYGAPQLQVEKVRTNDRKELGEVRVQYTGRDSFKAFAKALGVMDDLKSGVPRAGYRGIVTFQFRGRRVHLAPPLTWEGYDPSWN

Specifications

Antigen MGAT1
Applications ELISA
Classification Monoclonal
Clone 2G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MGAT1.
Formulation ascites with no preservative
Gene MGAT1
Gene Accession No. NM_002406
Gene Alias GLCNAC-TI/GLCT1/GLYT1/GNT-1/GNT-I/MGAT
Gene Symbols MGAT1
Host Species Mouse
Immunogen MGAT1 (NP_002397,348 a.a. ∽ 445 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4245
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.