Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGC50273, Mouse, Clone: 3B10, Abnova™

Catalog No. 89107802 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-802 Supplier Abnova Corporation Supplier No. H00408029M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MGC50273.

Sequence: MFVRPESGEQGPETLAPASGAEIQRFPVPAVEPVPAPGADSPPGTALELEEAPEPSCRCPGTAQDQPSEELPDFMAPPVEPPASALELKVWLELEVAERGGQHSSSQQLPHCSQSWAQWKLWRQRPGFAIWAPLPHWRGTSLIQQSSSPAAEGPAATAAGAVCLPAGGAGEQEKEPVSRGSSRSSCSQRRPPPPGMEVCPQLGIWAICP

Specifications

Antigen MGC50273
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MGC50273.
Formulation PBS with no preservative; pH 7.4
Gene MGC50273
Gene Accession No. NM_214461.1
Gene Symbols MGC50273
Host Species Mouse
Immunogen MGC50273 (NP_999626.1,1 a.a. ∽ 209 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408029
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.