Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGMT, Mouse, Clone: MT 23.2, Abnova™

Catalog No. 89110978 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-978 Supplier Abnova Corporation Supplier No. MAB2383
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MGMT.

Specifications

Antigen MGMT
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MT 23.2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MGMT.
Dilution Flow Cytometry (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene MGMT
Gene Symbols MGMT
Host Species Mouse
Immunogen Recombinant protein corresponding to human MGMT.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4255
Test Specificity This antibody is specific to MGMT.
Target Species Human
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.