Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II I Ad, Mouse, Clone: 34-5-3, Abnova™
Mouse monoclonal antibody raised against MHC Class II I Ad.
Supplier: Abnova Corporation MAB8225
Specifications
MHC Class II I Ad | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (1 ug/106 cells The optimal working dilution should be determined by the end user. | |
Mouse | |
500 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry | |
34-5-3 | |
Mouse monoclonal antibody raised against MHC Class II I Ad. | |
In 100mM BBS, pH 8.2 | |
(C57BL/6 x DBA/2) F1 mouse splenocytes. | |
RUO | |
It cross reacts with I-Ab and with cells from mice of the H-2p and H-2q halotypes. | |
Store at 4°C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction