Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIF, Mouse, Clone: 987s, Abnova™

Catalog No. 89113608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-608 Supplier Abnova Corporation Supplier No. MAB8836
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MIF.

This gene encodes a lymphokine involved in cell-mediated immunity, immunoregulation, and inflammation. It plays a role in the regulation of macrophage function in host defense through the suppression of anti-inflammatory effects of glucocorticoids. This lymphokine and the JAB1 protein form a complex in the cytosol near the peripheral plasma membrane, which may indicate an additional role in integrin signaling pathways. [provided by RefSeq

Specifications

Antigen MIF
Applications ELISA, Western Blot
Classification Monoclonal
Clone 987s
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MIF.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene MIF
Gene Alias GIF/GLIF/MMIF
Gene Symbols MIF
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-195 of human BCL2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4282
Test Specificity This antibody recognizes recombinant protein MIF.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.