Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MITF, Mouse, Clone: C5, Abnova™

Catalog No. 89110725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-725 Supplier Abnova Corporation Supplier No. MAB1955
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MITF.

This gene encodes a transcription factor that contains both basic helix-loop-helix and leucine zipper structural features. It regulates the differentiation and development of melanocytes retinal pigment epithelium and is also responsible for pigment cell-specific transcription of the melanogenesis enzyme genes. Heterozygous mutations in the this gene cause auditory-pigmentary syndromes, such as Waardenburg syndrome type 2 and Tietz syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen MITF
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MITF.
Dilution Western blot (1 ug/mL) Immunoprecipitation (2 ug/mg of protein lysate) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene MITF
Gene Alias MI/WS2A/bHLHe32
Gene Symbols MITF
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus of human MITF.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4286
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.