Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKI67, Mouse, Clone: MIB-1, Abnova™

Catalog No. 89111464 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-464 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-464 Supplier Abnova Corporation Supplier No. MAB3545
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MKI67.

This gene encodes a nuclear protein that is associated with and may be necessary for cellular proliferation. Alternatively spliced transcript variants have been described. A related pseudogene exists on chromosome X. [provided by RefSeq

Specifications

Antigen MKI67
Applications Immunohistochemistry
Classification Monoclonal
Clone MIB-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MKI67.
Dilution The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer, pH 7.3 (0.09% sodium azide)
Gene MKI67
Gene Alias KIA/Ki-67
Gene Symbols MKI67
Host Species Mouse
Immunogen Native purified MKI67 from human nuclear Ki-67 antigen.
Quantity 5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4288
Test Specificity This antibody reacts with MKI67 nuclear protein.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.