Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKI67IP, Mouse, Clone: 18E148, Abnova™

Catalog No. 89107877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-877 Supplier Abnova Corporation Supplier No. MAB0110
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of MKI67IP.

This gene encodes a protein that interacts with the forkhead-associated domain of the Ki-67 antigen. The encoded protein may bind RNA and may play a role in mitosis and cell cycle progression. Multiple pseudogenes exist on chromosomes 5, 10, 12, 15, and 19.[provided by RefSeq

Specifications

Antigen MKI67IP
Applications Western Blot
Classification Monoclonal
Clone Clone: 18E148
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of MKI67IP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene MKI67IP
Gene Alias NIFK/Nopp34
Gene Symbols MKI67IP
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human MKI67IP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84365
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.