Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKX (C10orf48), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107742 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-742 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-742 Supplier Abnova Corporation Supplier No. H00283078A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant C10orf48.

MKX is a member of an Iroquois (IRX) family-related class of 'three-amino acid loop extension' (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain (Anderson et al., 2006 [PubMed 16408284]).[supplied by OMIM

Sequence: MNTIVFNKLSSAVLFEDGGASERERGGRPYSGVLDSPHARPEVGIPDGPPLKDNLGLRHRRTGARQNGGKVRHKRQALQDMARPLKQWLYKHRDN

Specifications

Antigen MKX (C10orf48)
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant C10orf48.
Formulation 50% glycerol
Gene MKX
Gene Accession No. NM_173576
Gene Alias C10orf48/IFRX/IRXL1/MGC39616
Gene Symbols MKX
Host Species Mouse
Immunogen C10orf48 (NP_775847, 1 a.a. ∽ 95 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283078
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.