Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLANA, Mouse, Clone: M2-7C10, Abnova™

Catalog No. 89109449 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-449 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-449 Supplier Abnova Corporation Supplier No. MAB11283
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MLANA.

Specifications

Antigen MLANA
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone M2-7C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MLANA.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500) The optimal working dilution should be determined by the end user.
Formulation In 0.2mg/mL PBS (0.05% BSA, 0.05% sodium azide)
Gene MLANA
Gene Alias MART-1/MART1
Gene Symbols MLANA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human MLANA.
Purification Method Protein G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.