Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-MLN64/STARD3, Clone: 2BE-2F4,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1283
Description
Specifically detects MLN64/STARD3 clone: 2BE-2F4 in Human samples, and it is validated for Immunohistochemistry, Western Blotting
MLN64/StARD3 (also known as es64, StAR-related lipid transfer protein 3, Metastatic lymph node gene 64 protein, Metastatic lymph node protein 64, StARD3, START domain-containing protein 3, Steroidogenic acute regulatory protein related; UniProt Q14849) is encoded by the MLN64 (or CAB1 & STARD3) gene in human (NCBI Gene ID 10948). MLN64/StARD3 is a late endosome (LE) multitransmembrane protein that plays an important role in intracellular cholesterol transport and steroidogenesis. In addition to binding cholesterol via its C-terminal STAR domain, MLN64/StARD3 is reported to interact with ER-anchored VAP proteins via a conserved motif called 'two phenylalanines in an acidic tract' or FFAT, thereby forming an LE-ER tethering complex at the inter-organelle membrane contacts sites (MCSs), allowing heterologous membrane apposition and the exchange of metabolites (PMID 23709693 and 24105263). Clone 2BE-2F4 is also known as 2BE2F4 and mAbMLN64-Ct, it detects an epitope within the c-terminal region encompassing the last 20% of MLN64 sequence.
Specifications
MLN64/STARD3 | |
Monoclonal | |
Mouse monoclonal IgG1κ ascites with 0.05% sodium azide. | |
STARD3; CAB1; MLN64 | |
Ovalbumin-conjugated linear peptide corresponding to human MLN64/STARD3 near the C-terminus. | |
100 μL | |
Signaling | |
NP_001159409 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
2BE-2F4 | |
Q14849 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction