Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MME, Mouse, Clone: MEM-78, Abnova™

Catalog No. 89108206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-206 Supplier Abnova Corporation Supplier No. MAB0875
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native MME.

This gene encodes a common acute lymphocytic leukemia antigen that is an important cell surface marker in the diagnosis of human acute lymphocytic leukemia (ALL). This protein is present on leukemic cells of pre-B phenotype, which represent 85% of cases of ALL. This protein is not restricted to leukemic cells, however, and is found on a variety of normal tissues. It is a glycoprotein that is particularly abundant in kidney, where it is present on the brush border of proximal tubules and on glomerular epithelium. The protein is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon, enkephalins, substance P, neurotensin, oxytocin, and bradykinin. This gene, which encodes a 100kDa type II transmembrane glycoprotein, exists in a single copy of greater than 45kb. The 5' untranslated region of this gene is alternatively spliced, resulting in four separate mRNA transcripts. The coding region is not affected by alternative splicing. [provided by RefSeq]

Specifications

Antigen MME
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM-78
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native MME.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene MME
Gene Alias CALLA/CD10/DKFZp686O16152/MGC126681/MGC126707/NEP
Gene Symbols MME
Host Species Mouse
Immunogen Native purified MME from human pre-B cell line NALM-6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4311
Test Specificity This antibody reacts with CD10 antigen (CALLA - Common acute lymphatic leukemia antigen), a 100 KDa type II integral membrane protein.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.