Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mme, Mouse, FITC, Clone: LT10, Abnova™

Catalog No. 89111770 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-770 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-770 Supplier Abnova Corporation Supplier No. MAB4348
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Mme.

Specifications

Antigen Mme
Applications Flow Cytometry
Classification Monoclonal
Clone LT10
Conjugate FITC
Description Mouse monoclonal antibody raised against native Mme.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene Mme
Gene Alias 6030454K05Rik/C85356/CALLA/CD10/NEP
Gene Symbols Mme
Host Species Mouse
Immunogen Mative purified Mme from mouse NALM-6 leukemia pre-B cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17380
Test Specificity This antibody reacts with CD10 antigen (CALLA - Common acute lymphatic leukemia antigen), a 100 KDa type II integral membrane protein.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.