Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP2, Mouse, Clone: SB13a, Abnova™

Catalog No. 89111351 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-351 Supplier Abnova Corporation Supplier No. MAB3330
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MMP2.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.[provided by RefSeq

Specifications

Antigen MMP2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SB13a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MMP2.
Dilution ELISA (1:3000-1:8000) Immunohistochemistry (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene MMP2
Gene Alias CLG4/CLG4A/MMP-II/MONA/TBE-1
Gene Symbols MMP2
Host Species Mouse
Immunogen Recombinant protein corresponding to human MMP2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4313
Test Specificity human MMP-2, Does not cross react to human MMP-1, MMP-3 or MMP-9.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.