Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP26, Mouse, Clone: 3B9, Abnova™

Catalog No. 89107136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-136 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-136 Supplier Abnova Corporation Supplier No. H00056547M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MMP26.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The encoded protein degrades type IV collagen, fibronectin, fibrinogen, casein, vitronectin, alpha 1-antitrypsin, alpha 2-macroglobulin, and insulin-like growth factor-binding protein 1, and activates MMP9 by cleavage. The protein differs from most MMP family members in that it lacks a conserved C-terminal protein domain. [provided by RefSeq

Sequence: SFWQWAHEDGWPFDGPGGILGHAFLPNSGNPGVVHFDKNEHWSASDTGYNLFLVATHEIGHSLGLQHSGNQSSIMYPTYWYHDPRTFQLSADDIQRIQHLYGEKCSSDIP

Specifications

Antigen MMP26
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MMP26.
Formulation ascites with no preservative
Gene MMP26
Gene Accession No. NM_021801
Gene Alias MGC126590/MGC126592
Gene Symbols MMP26
Host Species Mouse
Immunogen MMP26 (NP_068573,152 a.a. ∽ 261 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56547
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.