Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP9, Mouse, Clone: 2C3, Abnova™

Catalog No. 89110723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-723 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-723 Supplier Abnova Corporation Supplier No. MAB1952
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of MMP9.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. [provided by RefSeq

Specifications

Antigen MMP9
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of MMP9.
Dilution Western blot (1 to 2 ug/mL) Immunohistochemistry on formalin to fixed/paraffin to embedded tissues (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene MMP9
Gene Alias CLG4B/GELB/MMP-9
Gene Symbols MMP9
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human MMP9.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4318
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.