Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Granulocyte, Mouse, Clone: 74-22-15, Abnova™

Catalog No. 89136153 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-153 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-153 Supplier Abnova Corporation Supplier No. MAB6219
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Monocyte/Granulocyte.

Specifications

Antigen Monocyte/Granulocyte
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 74-22-15
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Monocyte/Granulocyte.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Mouse
Immunogen dd Miniature swine thymocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Porcine SWC3a.
Target Species Pig
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.