Learn More
Specifications
Specifications
| Antigen | Morphine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | BDI918 |
| Conjugate | Unconjugated |
| Formulation | In PBS, pH 7.2 (0.05% sodium azide) |
| Host Species | Mouse |
| Immunogen | Morphine conjugated with BTG. |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.