Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD157 Mouse anti-Mouse, Brilliant Violet 421, Clone: BP-3, BD Optibuild™

Catalog No. BDB740085 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740085 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740085 Supplier BD Biosciences Supplier No. 740085
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BP-3 antibody reacts with CD157, also known as the BP-3 alloantigen or Bone marrow ST romal cell antigen- 1 (BST-1), found in a variety of common laboratory strains of mice (AKR, BALB/c, CBA/N, C3H/HeJ, C57BL/6, DBA/2, NZB, C.B-17, SJL and Swiss, but not A/J). CD157 is a differentially glycosylated GPI-linked cell-surface glycoprotein with a core protein of 32 kDa and differing patterns of glycosylation in different types of cells. It is a member, along with CD38, of a family of genetically-related ectoenzymes. The BP-3 alloantigen is expressed on the B lineage from the earliest detectable progenitors to circulating immature B lymphocytes, on immature thymocytes up to the expression of the T-cell receptor, on circulating neutrophils, peritoneal macrophages, on subpopulations of stromal cells in peripheral lymphoid organs (but not in bone marrow or thymus), on intestinal epithelial cells, and in the lumen of kidney-collecting tubules. The distribution of CD157 antigen on leukocytes and stromal cells of lymphoid organs suggests that it may be involved in leukocyte-stroma interactions which support the development, migration, and/or responses of leukocyte populations.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD157
Applications Flow Cytometry
Classification Monoclonal
Clone BP-3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description BP-3 Alloantigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse pre-B cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.