Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD212 Mouse anti-Mouse, Brilliant Violet 421, Clone: 114, BD Optibuild™

Catalog No. BDB740051 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740051 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740051 Supplier BD Biosciences Supplier No. 740051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 114 monoclonal antibody specifically binds to mouse CD212 (the β 1 subunit of IL-12R β 1), originally termed IL-12R β , of the mouse IL-12 receptor complex. The IL-12R β 1 subunit associates with a β 2 subunit to form a heterodimeric IL-12 receptor complex. Each one of the IL-12R subunits exhibits low affinity for IL-12, but in combination, they bind IL-12 with high affinity. The IL-12R β 1 subunit interacts primarily with IL-12 p40 whereas the IL-12R β β 1 has more recently been described to bind IL-23, a heterodimer formed of the p40 subunit from IL-12, and p19. The cytoplasmic regions of the β 1 and β 2 subunits contain the box1 and box2 motifs found in other cytokine receptors such as gp130, LIFR and G-CSFR. IL-12R β 1 are primarily expressed by activated T cells and NK cells. Experiments with IL-12R β 1 deficient mice have shown that IL-12R β 1 is necessary for mouse T and NK cell responsiveness to IL-12 p75. The 114 antibody was generated by immunizing IL-12R β 1 deficient mice of (129 x BALB/c)F1 background with mouse Ba/F3 cells that were stably transfected with IL-12R β 1

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD212
Applications Flow Cytometry
Classification Monoclonal
Clone 114
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Il12rb1; IL-12R-beta-1; IL-12 Receptor β1 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IL-12Rβ1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.