Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD244.2 Mouse anti-Mouse, Brilliant Violet 605, Clone: 2B4, BD Optibuild™

Catalog No. BDB740345 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740345 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740345 Supplier BD Biosciences Supplier No. 740345
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD244 is a type I transmembrane glycoprotein that is also known as Natural killer cell receptor 2B4 (NKR2B4), or Signaling lymphocytic activation molecule 4 (SLAMF4). CD244 is a member of the CD2 subset of the immunoglobulin superfamily (CD2 IgSF). It is expressed on all natural killer (NK) cells, IL-2-activated NK (LAK) cells, committed progenitors of NK cells, and a subset of T lymphocytes which mediate non-MHC-restricted cytoxicity, including dendritic epidermal T cells. The 2B4 monoclonal antibody specifically binds to CD244.2, the 2B4 alloantigen which is expressed on C57BL/6 and C58/J mice, but not in most strains tested (A/J, AKR, BALB/c, CBA/J, CBA/N, C3H/He, C57BR, DBA/1, DBA/2, NZB, SJL/J, 129). At least two isoforms of CD244.2 protein, products of alternative splicing of hnRNA, are expressed on IL-2-activated C57BL/6 NK cells. They differ only in their cytoplasmic domains; with 2B4L (150-aa cytoplasmic tail) having inhibitory activity and 2B4S (93-aa tail) being stimulatory. The extracellular domain of CD244 is a ligand for another CD2 IgSF member, CD48. 2B4 antibody activates lytic and secretory functions of IL-2-cultured NK cells and CD244.2+ T cells.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD244.2
Applications Flow Cytometry
Classification Monoclonal
Clone 2B4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description CD242.2; 2B4 B6 Alloantigen; Ly90; NAIL; Nmrk; SLAMF4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen rIL-2-propagated NK1.1+ cells from C57BL/6 mice
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.