Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP2, Mouse, Clone: 2B9, Abnova™

Catalog No. 89104939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-939 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-939 Supplier Abnova Corporation Supplier No. H00004355M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MPP2.

Palmitoylated membrane protein 2 is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. Palmitoylated membrane protein 2 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. [provided by RefSeq

Sequence: MPVAATNSETAMQQVLDNLGSLPSATGAAELDLIFLRGIMESPIVRSLAKAHERLEETKLEAVRDNNLELVQEILRDLAQLAEQSSTAAELAHILQEPHFQSLLETHDSVASKTYETPPPSPGLDPTFSNQPVPPDAVRMVGIRKTAGEHLGVTFRVEGGELVIARILHGGMVAQQGLLHVGDIIKEVNGQPVGSDPRALQELLRNASGSVILKILPSYQEPHLPRQVFVKCHFDYDPARDSLIPCKEAGLRFNAGDLLQIVNQDDANWWQACHVEGGSAGLIPSQLLEEKRKAFVKRDLELTPNSGTLCGSLSGKKKKRMMYLTTKNAEFDRHELLIYEEVARMPPFRRKTLVLIGAQGVGRRSLKNKLIMWDPDRYGTTVPYTSRRPKDSEREGQGYSFVSRGEMEADVRAGRYLEHGEYEGNLYGTRIDSIRGVVAAGKVCVLDVNPQAVKVLRTAEFVPYVVFIEAPDFETLRAMNRAALESGISTKQLTEADLRRTVEESSRIQRGYGHYFDLCLVNSNLERTFRELQTAMEKLRTEPQWVPVSWVY

Specifications

Antigen MPP2
Applications ELISA
Classification Monoclonal
Clone 2B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MPP2.
Formulation ascites with no preservative
Gene MPP2
Gene Accession No. BC030287
Gene Alias DKFZp686A06252/DKFZp686J2189/DKFZp761D0712/DLG2
Gene Symbols MPP2
Host Species Mouse
Immunogen MPP2 (AAH30287,1 a.a. ∽ 552 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4355
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.