Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-MPR300/IGF-2R/CD222, Clone: 2C2,

Catalog No. MABF935 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF935 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF935 Supplier MilliporeSigma™ Supplier No. MABF935
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MPR300/IGF-2R/CD222 clone: 2C2 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting

Cation-independent mannose-6-phosphate receptor (UniProt P11717; also known as 300kDa mannose 6-phosphate receptor, CD222, CI Man-6-P receptor, CI-MPR, IGF-II receptor, Insulin-like growth factor 2 receptor, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6P/IGF2R, M6P-R, M6PR, MPR 300) is encoded by the IGF2R (also known as CIMPR, MPRI) gene (Gene ID 3482) in human. Mannose 6-phosphate (M6P) receptors (MPRs) mediate the delivery of newly synthesized, mannose 6-phosphate-bound lysosomal hydrolases from the secretory pathway to the endosomal system. MPRs bind lysosomal hydrolases in the trans-Golgi network (TGN) and unload the enzyme in the endosomal system. There exist a cation-independent CI-MPR (IGF2R) and a smaller CD-MPR that requires divalent cations for lysosomal hydrolases recognition. MPRs also undergo recycling between the endosomal system and the plasma membrane, where CI-MPR mediates the internalization of other ligands, including insulin-like growth factor II (IGF-II). CI-MPR/IGF2R is produced with a signal peptide (a.a. 1-40), the removal of which yields the mature receptor with a large lumenal/extracellular region (a.a. 41-2304), a transmembrane segment (a.a. 2305-2327), and a cytoplasmic domain (a.a. 2328-2491), The extracellular region is composed of 15 P-type carbohydrate recognition domains, also referred to as MRH (mannose 6-phosphate receptor homology) domains. Domains 3 and 9 bind M6P with high affinity and domain 5 binds M6P with weak affinity, while domain 11 binds IGF-II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPR300/IGF-2R/CD222
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2C2
Formulation Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.
Gene Accession No. P11717
Gene Symbols IGF2R; CIMPR; MPRI
Host Species Mouse
Immunogen Recombinant full-length human MPR300/IGF-2R/CD222.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000867
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.