Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRRF, Mouse, Clone: AT7D10, Abnova™

Catalog No. 89109372 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-372 Supplier Abnova Corporation Supplier No. MAB11203
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MRRF.

Specifications

Antigen MRRF
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT7D10
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MRRF.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.01% Sodium azide
Gene MRRF
Gene Alias MRFF/MTRRF/RRF
Gene Symbols MRRF
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 56-262 of human MRRF.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.