Learn More
MS4A1, Mouse, APC-Cy7, Clone: B-Ly1, Abnova™
Mouse monoclonal antibody raised against MS4A1.
Supplier: Abnova Corporation MAB5996
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. [provided by RefSeq
Specifications
MS4A1 | |
Monoclonal | |
APC-Cyanine7 | |
Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
MS4A1 | |
MS4A1 | |
Human MS4A1. | |
RUO | |
931 | |
Human | |
Liquid |
Flow Cytometry, Immunoprecipitation | |
B-Ly1 | |
Mouse monoclonal antibody raised against MS4A1. | |
In PBS (0.09% sodium azide) | |
B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7 | |
Mouse | |
100 Reactions | |
Primary | |
Specificity human CD20 | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.