Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A1, Mouse, Clone: BC1, Abnova™

Catalog No. 89112023 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-023 Supplier Abnova Corporation Supplier No. MAB4973
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MS4A1.

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. [provided by RefSeq

Specifications

Antigen MS4A1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone BC1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MS4A1.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide)
Gene MS4A1
Gene Alias B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7
Gene Symbols MS4A1
Host Species Mouse
Immunogen Normal human lymphocytes from lymph node.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 931
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.