Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI2, Mouse, Clone: 2H4, Abnova™

Catalog No. 89107589 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-589 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-589 Supplier Abnova Corporation Supplier No. H00124540M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MSI2.

This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MEANGSQGTSGSANDSQHDPGKMFIGGLSWQTSPDSLRDYFSKFGEIRECMVMRDPTTKRSRGFGFVTFADPASVDKVLGQPHHELDSKTIDPKVAFPRRAQPKMVTRTKKIFVGGLSANTVVEDVKQYFEQFGKVEDAMLMFDKTTNRHRGFGFVTFENEDVVEKVCEIHFHEINNKMVECKKAQPKEVMFPPGTRGRARGLPYTMDAFMLGMGMLGYPNFVATYGRGYPGFAPSYGYQFPGFPAAAYGPVAAAAVAAARGSGSNPARPGGFPGANSPGPVADLYGPASQDSGVGNYISAASPQPGSGFGHGIAGPLIATAFTNGYH

Specifications

Antigen MSI2
Applications ELISA
Classification Monoclonal
Clone 2H4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MSI2.
Formulation ascites with no preservative
Gene MSI2
Gene Accession No. BC001526
Gene Alias FLJ36569/MGC3245/MSI2H
Gene Symbols MSI2
Host Species Mouse
Immunogen MSI2 (AAH01526,1 a.a. ∽ 328 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124540
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.