Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSLN, Mouse, Clone: MN-1, Abnova™

Catalog No. 89110747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-747 Supplier Abnova Corporation Supplier No. MAB1993
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MSLN.

An antibody that reacts with ovarian cancers and mesotheliomas was used to isolate a cell surface antigen named mesothelin. Although the function of mesothelin is unknown, it may play a role in cellular adhesion and is present on mesothelium, mesotheliomas, and ovarian cancers. This gene encodes a preproprotein that is cleaved into two products, megakaryocyte potentiating factor and mesothelin. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen MSLN
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MN-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MSLN.
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MSLN
Gene Alias CAK1/MPF/SMR
Gene Symbols MSLN
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of human MSLN.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10232
Test Specificity This antibody is directed against human mesothelin protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.