Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSMB, Mouse, Clone: YPSP-4, Abnova™

Catalog No. 89112835 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-835 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-835 Supplier Abnova Corporation Supplier No. MAB6369
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MSMB.

The protein encoded by this gene is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq

Specifications

Antigen MSMB
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone YPSP-4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MSMB.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene MSMB
Gene Alias HPC13/IGBF/MSP/MSPB/PN44/PRPS/PSP/PSP-94/PSP57/PSP94
Gene Symbols MSMB
Host Species Mouse
Immunogen Purified MSMB.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4477
Test Specificity This antibody react with MSMB or human prostate epithelial cells.
Target Species Human
Content And Storage Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.