Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTDH, Mouse, Clone: 2F11C3, Abnova™

Catalog No. 89109019 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-019 Supplier Abnova Corporation Supplier No. MAB10817
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MTDH.

Specifications

Antigen MTDH
Applications ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 2F11C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MTDH.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.03% sodium azide)
Gene MTDH
Gene Alias 3D3/AEG-1/AEG1/LYRIC
Gene Symbols MTDH
Host Species Mouse
Immunogen Recombinant protein corresponding to human MTDH.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92140
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.