Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRAP1, Mouse, Clone: 215Q18, Abnova™

Catalog No. 89110728 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-728 Supplier Abnova Corporation Supplier No. MAB1958

Mouse monoclonal antibody raised against partial recombinant FRAP1.

The protein encoded by this gene belongs to a family of phosphatidylinositol kinase-related kinases. These kinases mediate cellular responses to stresses such as DNA damage and nutrient deprivation. This protein acts as the target for the cell-cycle arrest and immunosuppressive effects of the FKBP12-rapamycin complex. The ANGPTL7 gene is located in an intron of this gene. [provided by RefSeq

Specifications

Antigen FRAP1
Applications Western Blot
Classification Monoclonal
Clone 215Q18
Conjugate Unconjugated
Description mechanistic target of rapamycin
Formulation In PBS, pH 7.3 (1% BSA, 0.08% sodium azide)
Gene MTOR
Gene Alias FRAP, FRAP1, FRAP2, RAFT1, RAPT1
Gene Symbols MTOR
Host Species Mouse
Immunogen Recombinant protein of human FRAP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.