Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-MTSS1, Clone: 15C3.2,

Catalog No. MABC607MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC607MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC607MI Supplier MilliporeSigma™ Supplier No. MABC607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MTSS1 clone: 15C3.2 in Human, Monkey samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Metastasis suppressor protein 1 (UniProt O43312; also known as Metastasis suppressor YGL-1, Missing in metastasis protein) is encoded by the MTSS1 (also known as KIAA0429, MIM) gene (Gene ID 9788) in human. MTSS1 is a scaffold protein that regulates cytoskeletal dynamics and actin polymerization. MTSS1 is also reported to bind Gli proteins or the Gli-Sufu complex and potentiate the transcription of effector genes of the sonic hedgehog signaling pathway. MTSS1 is down-regulated in many metastatic cancer cells compared with their non-metastatic counterparts. MTSS1 expression is shown to be inversely correlated with the prognosis of solid carcinomas of the breast, liver, and bladder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTSS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 15C3.2
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O43312
Gene Symbols MTSS1; KIAA0429; MIM
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal of human MTSS1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_055566
Target Species Human, Monkey
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.