Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTUS1, Mouse, Clone: 1C10, Abnova™

Catalog No. 89107193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-193 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-193 Supplier Abnova Corporation Supplier No. H00057509M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MTUS1.

This gene encodes a protein which contains a C-terminal domain able to interact with the angiotension II (AT2) receptor and a large coiled-coil region allowing dimerization. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. One of the transcript variants has been shown to encode a mitochondrial protein that acts as a tumor suppressor and partcipates in AT2 signaling pathways. Other variants may encode nuclear or transmembrane proteins but it has not been determined whether they also participate in AT2 signaling pathways. [provided by RefSeq

Sequence: MQLQEQFDNLNAAHETSKLEIEASHSEKLELLTKAYEASLSEIKKGHEIEKKSLEDLLSEKQESLEKQINDLKSENDALNEKLKSEEQKRRAREKANLKNPQIMYLEQELESLKAVLEIKNEKLHQQDIKLMKMGKLVDNNTALVDKLKRFQQENEELKARMDKHMAISRQLSTEQAVLQESLEKESKVNKRLSMENEELLWKLHNGDLCSPKRSPTSSAIPLQSPRNSGSFPSPSISPR

Specifications

Antigen MTUS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MTUS1.
Formulation ascites with no preservative
Gene MTUS1
Gene Accession No. BC033842
Gene Alias ATIP/DKFZp586D1519/DKFZp686F20243/FLJ14295/KIAA1288/MP44/MTSG1
Gene Symbols MTUS1
Host Species Mouse
Immunogen MTUS1 (AAH33842,1 a.a. ∽ 240 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57509
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.