Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC16, Mouse, Clone: 110, Abnova™

Catalog No. 89112827 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-827 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-827 Supplier Abnova Corporation Supplier No. MAB6357
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MUC16.

Specifications

Antigen MUC16
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MUC16.
Dilution ELISA (0.125 ng/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene MUC16
Gene Alias CA125/FLJ14303
Gene Symbols MUC16
Host Species Mouse
Immunogen Human MUC16 protein.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94025
Test Specificity This antibody reacts with human CA 125.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.