Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-MUC16, Clone: 23, Abnova™

Catalog No. 89113353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-353 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-353 Supplier Abnova Corporation Supplier No. MAB8105
Add to Cart
Add to Cart

Specifications

Applications ELISA
Classification Monoclonal
Clone 23
Description Mouse monoclonal antibody raised against native MUC16.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene Alias CA125/FLJ14303
Gene Symbols MUC16
Immunogen Native purified human MUC16.
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) 94025
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.