Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2, Mouse, Clone: CCP58, Abnova™

Catalog No. 89109487 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-487 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-487 Supplier Abnova Corporation Supplier No. MAB11323
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MUC2.

This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq

Specifications

Antigen MUC2
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CCP58
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MUC2.
Dilution Western Blot (1:100-1:2000) Flow Cytometry (1:10 - 1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200 - 1:400) Immunoprecipitation (1:10-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene MUC2
Gene Alias MLP/SMUC
Gene Symbols MUC2
Host Species Mouse
Immunogen The 23 amino acid VNTR region of human MUC2. This antibody is known to be specific to amino acids GTQTP.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4583
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.