Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-954 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-954 Supplier Abnova Corporation Supplier No. H00004585A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MUC4.

The major constituents of mucus, the viscous secretion that covers epithelial surfaces such as those in the trachea, colon, and cervix, are highly glycosylated proteins called mucins. These glycoproteins play important roles in the protection of the epithelial cells and have been implicated in epithelial renewal and differentiation. This gene encodes an integral membrane glycoprotein found on the cell surface, although secreted isoforms may exist. At least two dozen transcript variants of this gene have been found, although for many of them the full-length transcript has not been determined or they are found only in tumor tissues. This gene contains a region in the coding sequence which has a variable number (>100) of 48 nt tandem repeats. [provided by RefSeq

Sequence: GVSLFPYGADAGDLEFVRRTVDFTSPLFKPATGFPLGSSLRDSLYFTDNGQIIFPESDYQIFSYPNPLPTGFTGRDPVALVAPFWDDADFSTGRGTTFYQEYETFYGEHS

Specifications

Antigen MUC4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MUC4.
Formulation 50% glycerol
Gene MUC4
Gene Accession No. NM_004532
Gene Alias HSA276359
Gene Symbols MUC4
Host Species Mouse
Immunogen MUC4 (NP_004523, 79 a.a. ∽ 188 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4585
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.