Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5AC, Mouse, Clone: SPM297, Abnova™

Catalog No. 89110257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-257 Supplier Abnova Corporation Supplier No. MAB12114
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MUC5AC.

Specifications

Antigen MUC5AC
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SPM297
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MUC5AC.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene MUC5AC
Gene Alias MUC5
Gene Symbols MUC5AC
Host Species Mouse
Immunogen M1 mucin prepared from the fluid of human ovarian mucinous cyst.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4586
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.