Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5B, Mouse, Clone: 8C11, Abnova™

Catalog No. 89107810 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-810 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-810 Supplier Abnova Corporation Supplier No. H00727897M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MUC5B.

Mucins are high molecular mass, highly glycosylated macromolecules that are the major components of mucus secretions. MUC5B is a salivary mucin that is thought to contribute to the lubricating and viscoelastic properties of whole saliva. It is composed of 14.9% protein, 78.1% carbohydrate, and 7% sulfate (Troxler et al., 1995 [PubMed 8554565]).[supplied by OMIM

Sequence: CEEDSCQVRINTTILWHQGCETEVNITFCEGSCPGASKYSAEAQAMQHQCTCCQERRVHEETVPLHCPNGSAILHTYTHVDECGCTPFCVPAPMAPPHTRGFPAQEATAV

Specifications

Antigen MUC5B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MUC5B.
Formulation ascites with no preservative
Gene MUC5B
Gene Accession No. XM_039877
Gene Alias MG1/MUC5/MUC9
Gene Symbols MUC5B
Host Species Mouse
Immunogen MUC5B (XP_039877.9,4186 a.a. ∽ 4295 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727897
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.