Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC7, Mouse, Clone: 7F2, Abnova™

Catalog No. 89104957 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-957 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-957 Supplier Abnova Corporation Supplier No. H00004589M06A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MUC7.

This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene

Sequence: HQSPKSHFELPHYPGLLAHQKPFIRKSYKCLHKRCRPKLPPSPNNPPKFPNPHQPPKHPDKNSSVVNPTLVATTQIPSVTFPSASTKITTLPNVTFLPQN

Specifications

Antigen MUC7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MUC7.
Formulation ascites with no preservative
Gene MUC7
Gene Accession No. NM_152291
Gene Alias DKFZp686J03256/FLJ27047/MG2/MGC34772
Gene Symbols MUC7
Host Species Mouse
Immunogen MUC7 (NP_689504,36 a.a. ∽ 135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4589
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.